missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR alpha/NR1C1 Antibody (OTI1E8), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73567UV
This item is not returnable.
View return policy
Description
PPAR alpha/NR1C1 Monoclonal specifically detects PPAR alpha/NR1C1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| PPAR alpha/NR1C1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PPARA | |
| Recombinant protein fragment corresponding to amino acids 145-420 of human PPAR alpha/NR1C1 (NP_005027) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1E8 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452 | |
| Mouse | |
| Affinity Purified | |
| Cancer, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Transcription Factors and Regulators | |
| 5465.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction