missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPAR alpha/NR1C1 Transcription Factor Assay Kit (Colorimetric)
Assay Kit (Colorimetric)
Brand: Novus Biologicals™ KA1356
This item is not returnable.
View return policy
Description

Specifications
Transcription Factor Assay Kit (Colorimetric) | |
Functional | |
PPARA | |
Cancer, Lipid and Metabolism, mTOR Pathway, Transcription Factors and Regulators | |
alpha, hPPAR, NR1C1MGC2237, Nuclear receptor subfamily 1 group C member 1, peroxisome proliferator-activated receptor alpha, PPARalpha, PPAR-alpha, PPARMGC2452 |
Nuclear Extracts | |
5465 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction