missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR delta/NR1C2 Antibody (PCRP-PPARD-4C2) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-23996JF525
This item is not returnable.
View return policy
Description
PPAR delta/NR1C2 Monoclonal antibody specifically detects PPAR delta/NR1C2 in Human samples. It is validated for Flow Cytometry
Specifications
| PPAR delta/NR1C2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| FAAR, MGC3931, NR1C2PPAR-delta, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB | |
| Recombinant fragment (around 72-146) of human PPAR delta/NR1C2 protein | |
| 0.1 mL | |
| Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators, Wnt Signaling Pathway | |
| 5467 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| PCRP-PPARD-4C2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction