missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPAR delta/NR1C2 Transcription Factor Assay Kit (Colorimetric)
Assay Kit (Colorimetric)
Brand: Novus Biologicals™ KA1359
This item is not returnable.
View return policy
Description

Specifications
Transcription Factor Assay Kit (Colorimetric) | |
Functional | |
PPARD | |
Diabetes Research, Lipid and Metabolism, Transcription Factors and Regulators, Wnt Signaling Pathway | |
FAAR, MGC3931, NR1C2PPAR-delta, NUC1PPAR-beta, NUCI, NUCII, Nuclear hormone receptor 1, Nuclear receptor subfamily 1 group C member 2, peroxisome proliferative activated receptor, delta, Peroxisome proliferator-activated receptor beta, peroxisome proliferator-activated receptor delta, PPARB |
Nuclear Extracts | |
5467 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction