missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPAR gamma Monoclonal Antibody (C5)

Product Code. 17824240
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17824240 200 μL 200µL
17664166 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17824240 Supplier Invitrogen™ Supplier No. MA541659

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPAR gamma Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR gamma
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene PPARG
Gene Accession No. O88275
Gene Alias CIMT1; GLM1; HUMPPARG; NR1C3; nuclear hormone receptor; nuclear hormone receptor; PPARG; Nuclear receptor subfamily 1 group C member 3; nuclear receptor transcription factor; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferative activated receptor, gamma; peroxisome proliferative activated receptor, gamma-2; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 1; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activated receptor gamma-1; peroxisome proliferator activated receptor gamma-2; peroxisome proliferator activatived receptor gamma; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; peroxisome proliferator-activated receptor; peroxisome proliferator-activated receptor gamma; peroxisome proliferator-activated receptor gamma 1; peroxisome proliferator-activated receptor gamma 1a; peroxisome proliferator-activated receptor gamma 1-a; peroxisome proliferator-activated receptor gamma 1b; peroxisome proliferator-activated receptor gamma 1-b; peroxisome proliferator-activated receptor gamma 1c; peroxisome proliferator-activated receptor gamma 1d; peroxisome proliferator-activated receptor gamma 2; peroxisome proliferator-activated receptor-gamma; peroxisome proliferator-activated receptor-gamma 2; peroxisome proliferator-cctivated receptor gamma 1; PPAR gamma; PPAR gamma 1; PPAR gamma 3; Pparg; PPARG1; PPARG2; PPARgamma; PPAR-gamma; PPARgamma2; PPAR-gamma2; PPARy; proliferator activated gamma; transcription factor
Gene Symbols PPARG
Host Species Mouse
Immunogen Recombinant protein Peroxisome Proliferator Activated Receptor Gamma. The antigen corresponds to amino acid range 311-493 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25664
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.