missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPAR gamma/NR1C3 Transcription Factor Assay Kit (Colorimetric)
Assay Kit (Colorimetric)
Brand: Novus Biologicals™ KA1358
This item is not returnable.
View return policy
Description

Specifications
Transcription Factor Assay Kit (Colorimetric) | |
Functional | |
PPARG | |
Cancer, mTOR Pathway | |
CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma |
Nuclear Extracts | |
5468 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction