Learn More
PPARA Mouse anti-Human, Mouse, Clone: 1331CT894.186.143, Abnova™
Mouse monoclonal antibody raised against human PPARA.
Brand: Abnova MAB12349.400uL
Description
Peroxisome proliferators include hypolipidemic drugs, herbicides, leukotriene antagonists, and plasticizers; this term arises because they induce an increase in the size and number of peroxisomes. Peroxisomes are subcellular organelles found in plants and animals that contain enzymes for respiration and for cholesterol and lipid metabolism. The action of peroxisome proliferators is thought to be mediated via specific receptors, called PPARs, which belong to the steroid hormone receptor superfamily. PPARs affect the expression of target genes involved in cell proliferation, cell differentiation and in immune and inflammation responses. Three closely related subtypes (alpha, beta/delta, and gamma) have been identified. This gene encodes the subtype PPAR-alpha, which is a nuclear transcription factor. Multiple alternatively spliced transcript variants have been described for this gene, although the full-length nature of only two has been determined. [provided by RefSeq]
Specifications
| PPARA | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (1:25) Immunohistochemistry (1:25) Immunofluorescence (1:25) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| MGC2237/MGC2452/NR1C1/PPAR/hPPAR | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 5465 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1331CT894.186.143 | |
| Mouse monoclonal antibody raised against human PPARA. | |
| In PBS (0.09% sodium azide) | |
| PPARA | |
| Recombinant protein corresponding to human PPARA. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.