missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPARA/PPARD/PPARG Complete Transcription Factor Assay Kit (Colorimetric)
Assay Kit (Colorimetric)
Brand: Novus Biologicals™ KA1357
This item is not returnable.
View return policy
Description

Specifications
Assay Kit (Colorimetric) | |
Functional | |
Storage of components varies. See protocol for specific instructions. |
Nuclear Extracts | |
1 Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction