missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPCDC Antibody (03), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06605IR
This item is not returnable.
View return policy
Description
PPCDC Monoclonal antibody specifically detects PPCDC in Human samples. It is validated for ELISA
Specifications
| PPCDC | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 03 | |
| ELISA | |
| coaC, EC 4.1.1.36, FLJ14585, MDS018, phosphopantothenoylcysteine decarboxylase, PPC-DC | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PPCDC (AAH14409.1; Met1-Ser204). | |
| 0.1 mL | |
| metabolism | |
| 60490 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction