Learn More
Invitrogen™ PPCS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588787
Description
Immunogen sequence: MAEMDPVAEF PQPPGAARWA EVMARFAARL GAQGRRVVLV TSGGTKVPLE ARPVRFLDNF SSGRRGATSA EAFLAAGYGV LFLYRARSAF PYAHRFPPQT WLSALRPSGP ALSGLLSLEA EENALPGFAE ALRSYQEAAA AGTFLAVEFT TLADYLHLLQ AAAQALNPLG PSAMFYLAAA VSDFYVPVSE MPEHKIQSSG GPLQITMKMV PKLLSPLVKD WAPKAFIISF KLETDPAIVI NRARKALEIY QHQVVVANIL ESRQSFVFIV TKDSETKLLL SEEEIEKGVE IEEKIVDNLQ SRHTAFIGDR N; Positive Samples: OVCAR-3, HeLa, HepG2.
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCS (EC 6.3.2.5), one of the last enzymes in this pathway, converts phosphopantothenate to phosphopantothenoylcysteine.
Specifications
| PPCS | |
| Polyclonal | |
| Unconjugated | |
| PPCS | |
| 6330579B17Rik; AI413471; AI790434; Coab; Phosphopantothenate--cysteine ligase; phosphopantothenoylcysteine synthetase; PPC synthetase; Ppcs; RGD1311399 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79717 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.53 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9HAB8 | |
| PPCS | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-311 of human PPCS (NP_0789402). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.