missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPFIA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560420
This item is not returnable.
View return policy
Description
Immunogen sequence: SSGHSTPRLA PPSPAREGTD KANHVPKEEA GAPRGEGPAI PGDTPPPTPR SARLERMTQA LALQAGSLED G Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 94%.
The PPFIA3 protein is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family, a family of transmembrane protein tyrosine phosphatases known to be important for axon guidance and mammary gland development. Similar to the other liprin family members, PPFIA3 can interact with ERC2, a member of the ELKS-Rab6-interacting protein-CAST family of proteins at the presynaptic termini, suggesting that these proteins may play a role in the molecular organization of presynaptic zones.
Specifications
| PPFIA3 | |
| Polyclonal | |
| Unconjugated | |
| PPFIA3 | |
| 2410127E16Rik; KIAA0654; liprin; liprin-alpha 3; Liprin-alpha3; liprin-alpha-3; liprin-alpha-3; LOW QUALITY PROTEIN: liprin-alpha-3; LPNA3; PPFIA3; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-3; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 3; protein tyrosine phosphatase, receptor type, f polypeptide, alpha 3; PTPRF interacting protein alpha 3; PTPRF-interacting protein alpha-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8541 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75145 | |
| PPFIA3 | |
| Recombinant protein corresponding to Human PPFIA3. Recombinant protein control fragment (Product #RP-99162). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction