missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPHLN1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 227.00 - € 326.00
Specifications
| Antigen | PPHLN1 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:500-1:2500, Immunoprecipitation 0.2-1 μL/mg of lysate |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18692042
|
Novus Biologicals
NBP2-98653-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18612885
|
Novus Biologicals
NBP2-98653-50ul |
50 μg |
€ 227.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPHLN1 Polyclonal antibody specifically detects PPHLN1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| PPHLN1 | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication | |
| PBS (pH 7.0) | |
| 51535 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:500-1:2500, Immunoprecipitation 0.2-1 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Gastric cancer antigen Ga50, HSPC232, MGC48786, periphilin 1, periphilin-1 | |
| Produced in rabbits immunized with E. coli-derived Human PPHLN1 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title