Learn More
PPID Mouse anti-Human, Unconjugated, Clone: 1A2B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67632-1-IG-150UL
Description
The PPID antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PPID. This antibody recognizes human, mouse, rat, pig antigen. The PPID antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PPID | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, ELISA | |
| 1A2B1 | |
| Unconjugated | |
| 150 μL | |
| 5481 | |
| IgG1 |