missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIF Monoclonal Antibody (OTI4D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809066S
This item is not returnable.
View return policy
Description
PPIF Monoclonal antibody specifically detects PPIF in Human samples. It is validated for Western BlotSpecifications
| PPIF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16860 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4879 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4D3 | |
| Unconjugated | |
| Nppb | |
| AW457192; cyclophilin 3; cyclophilin D; Cyclophilin F; CYP3; CypD; CyP-D; CyP-F; CyP-M; mitochondrial cyclophilin; mitochondrial Cyclophilin D; mitochondrial peptidyl-prolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase F; peptidyl-prolyl cis-trans isomerase F, mitochondrial; peptidyl-prolyl cis-trans isomerase, mitochondrial; peptidylprolyl isomerase F; peptidylprolyl isomerase F (cyclophilin F); PPIase; PPIase F; PPIF; Rotamase F | |
| Human recombinant protein fragment corresponding to amino acids 27-102 of human NPPB produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction