missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIF Monoclonal Antibody (OTI6E7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809037
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is part of the mitochondrial permeability transition pore in the inner mitochondrial membrane. Activation of this pore is thought to be involved in the induction of apoptotic and necrotic cell death.Specifications
| PPIF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P30405 | |
| PPIF | |
| Full length human recombit protein of human PPIF produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6E7 | |
| Unconjugated | |
| PPIF | |
| CYP3, CyP-M, Cyp-D, CypD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10105 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction