missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPIG Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105260
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PPIG.
Structural Maintenance of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregation defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.
Specifications
| PPIG | |
| Polyclonal | |
| Unconjugated | |
| PPIG | |
| AU019516; AU022200; B230312B02Rik; CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; CYP; Matrin cyclophilin; matrin cyclophilin (matrin-cyp); matrin-cyp; MGC133241; OTTHUMP00000204761; OTTHUMP00000204762; Peptidyl-prolyl cis-trans isomerase G; peptidylprolyl isomerase G; peptidyl-prolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); peptidyl-prolyl isomerase G (cyclophilin G); PPIase G; PPIG; Rotamase G; SCAF10; SR-cyclophilin; SRCyp; SR-cyp; SR-related CTD-associated factor 10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 228005, 83624, 9360 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2AR02, O55035, Q13427 | |
| PPIG | |
| A synthesized peptide derived from human PPIG(Accession Q13427), corresponding to amino acid residues R350-R400. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction