missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | PPIH |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227439
|
Novus Biologicals
NBP3-36707-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231889
|
Novus Biologicals
NBP3-36707-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PPIH Polyclonal antibody specifically detects PPIH in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PPIH | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol | |
| 10465 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| cyclophilin H, CYP20, CypH, CYPHpeptidyl prolyl isomerase H (cyclophilin H), EC 5.2.1.8, MGC5016, peptidyl-prolyl cis-trans isomerase H, peptidylprolyl isomerase H (cyclophilin H), PPIase H, Rotamase H, Small nuclear ribonucleoprotein particle-specific cyclophilin H, SnuCyp-20, USA-CyP SnuCyp-20, USA-CYPCYP-20, U-snRNP-associated cyclophilin SnuCyp-20, U-snRNP-associated cyclophilin SunCyp-20 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-177 of human PPIH (NP_006338.1).,, Sequence:, MAVANSSPVNPVVFFDVSIGGQEVGRMKIELFADVVPKTAENFRQFCTGEFRKDGVPIGYKGSTFHRVIKDFMIQGGDFVNGDGTGVASIYRGPFADENFKLRHSAPGLLSMANSGPSTNGCQFFITCSKCDWLDGKHVVFGKIIDGLLVMRKIENVPTGPNNKPKLPVVISQCGEM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title