missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL1 Monoclonal Antibody (OTI8F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803795
This item is not returnable.
View return policy
Description
This gene is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.Specifications
| PPIL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9Y3C6 | |
| PPIL1 | |
| Full length human recombit protein of human PPIL1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI8F8 | |
| Unconjugated | |
| PPIL1 | |
| CGI-124, CYPL1, PPIase, hCyPX | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51645 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction