missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPIL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557237
This item is not returnable.
View return policy
Description
Immunogen sequence: MENVESDPKT DRPKEEIRID ATTVFVDPYE EADAQIAQER KTQLKVAPET KVKSSQPQAG SQGPQTFRQG VGKYINPAAT KRAAEEEPST SATVPMSKKK PSRGFGDFSS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. This protein interacts with the proteinase inhibitor eglin c and is localized in the nucleus. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| PPIL2 | |
| Polyclonal | |
| Unconjugated | |
| PPIL2 | |
| 0610009L05Rik; 1700016N17Rik; 4921520K19Rik; 4930511F14Rik; AA589416; C130078A06Rik; CYC4; cyclophilin, 60kDa; cyclophilin-60; Cyclophilin-like protein Cyp-60; CYP60; Cyp-60; hCyP-60; peptidylprolyl cis-trans isomerase; peptidyl-prolyl cis-trans isomerase-like 2; peptidylprolyl isomerase (cyclophilin)-like 2; peptidylprolyl isomerase like 2; peptidylprolyl isomerase-like 2; PPIase; PPIL2; Probable inactive peptidyl-prolyl cis-trans isomerase-like 2; RING-type E3 ubiquitin-protein ligase PPIL2; Rotamase PPIL2; U-box domain containing 7; UBOX7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23759 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13356 | |
| PPIL2 | |
| Recombinant protein corresponding to Human PPIL2. Recombinant protein control fragment (Product #RP-95345). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction