missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPIL6 Monoclonal Antibody (OTI4F2)
GREENER_CHOICE

Product Code. 15995584
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15995584 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15995584 Supplier Invitrogen™ Supplier No. MA525992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PPIL6 Monoclonal Antibody for Western Blot, IHC (P)

PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIL6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PPIL6
Gene Accession No. Q8IXY8
Gene Alias 2900084F20Rik; bA425D10.6; Cyclophilin-like protein PPIL6; dJ919F19.1; peptidyl-prolyl cis-trans isomerase-like 6; peptidyl-prolyl cis-trans isomerase-like variant a; peptidylprolyl isomerase (cyclophilin)-like 6; peptidylprolyl isomerase like 6; PPIase; PPIL6; Probable inactive peptidyl-prolyl cis-trans isomerase-like 6; radial spoke 12 homolog; rotamase PPIL6; RSPH12
Gene Symbols PPIL6
Host Species Mouse
Immunogen Full length human recombinant protein of PPIL6 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285755
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.