missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL10 Monoclonal Antibody (OTI4H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800445S
This item is not returnable.
View return policy
Description
BCL10 Monoclonal antibody specifically detects BCL10 in Canine, Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| BCL10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A2A432, Q13620 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 492102, 72584, 8450 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4H9 | |
| Unconjugated | |
| CUL4B | |
| AI132454; B cell CLL/lymphoma 10; B cell leukemia/lymphoma 10; B-cell CLL/lymphoma 10; B-cell leukemia/lymphoma 10; B-cell lymphoma/leukemia 10; BCL10; Bcl-10; BCL10 immune signaling adaptor; C81403; CARD containing molecule enhancing NF-kB; CARD-containing apoptotic signaling protein; CARD-containing molecule enhancing NF-kappa-B; CARD-containing proapoptotic protein; CARD-like apoptotic protein; CARMEN; caspase-recruiting domain-containing protein; cCARMEN; cE10; c-E10; CED-3/ICH-1 prodomain homologous E10-like regulator; cellular homolog of vCARMEN; Cellular-E10; CIPER; Clap; hCLAP; IMD37; Mamm; mammalian CARD-containing adapter molecule E10; mCIPER; mCLAP; mE10; OTTHUMP00000011647; OTTHUMP00000036080; RCD; RP11-234D19.2; R-RCD1 | |
| Human recombinant protein fragment corresponding to amino acids 1-300 of human CUL4B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction