Learn More
PPM1B Mouse anti-Human, Clone: OTI2D9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503006
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional transcript variants have been described, but currently do not represent full-length sequences.Specifications
| PPM1B | |
| Monoclonal | |
| Unconjugated | |
| PPM1B | |
| PP2C-beta, PP2C-beta-X, PP2CB, PP2CBETA, PPC2BETAX | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5495 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI2D9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O75688 | |
| PPM1B | |
| Full length human recombit protein of human PPM1B produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.