missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1G Monoclonal Antibody (OTI1C12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502232
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.Specifications
| PPM1G | |
| Monoclonal | |
| Unconjugated | |
| PPM1G | |
| PP2CG, PP2CGAMMA, PPP2CG | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 475703, 5496 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1C12 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15355 | |
| PPM1G | |
| Full length human recombit protein of human PPM1G produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction