missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPM1G Recombinant Rabbit Monoclonal Antibody (JE64-91)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA544762

Product Code. 17828662

  • € 502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PPM1G Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways. This phosphatase is found to be responsible for the dephosphorylation of Pre-mRNA splicing factors, which is important for the formation of functional spliceosome. Studies of a similar gene in mice suggested a role of this phosphatase in regulating cell cycle progression. Alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPM1G
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
F1LNI5, O15355, Q61074
Ppm1g
Recombinant protein within human PPM1G aa 301-500/546.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
JE64-91
Unconjugated
Ppm1g
AI385652; fibroblast growth factor inducible 13; fibroblast growth factor-inducible protein 13; Fin13; PP2C, gamma; PP2CG; PP2CGAMMA; PP2C-gamma; Ppm1c; Ppm1g; PPP2CG; protein phosphatase 1C; Protein phosphatase 1G; protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform; protein phosphatase 2, catalytic subunit, gamma isoform; protein phosphatase 2C gamma isoform; protein phosphatase 2C isoform gamma; Protein phosphatase magnesium-dependent 1 gamma; protein phosphatase, Mg2+/Mn2+ dependent 1G; protein phosphatase, Mg2+/Mn2+ dependent, 1G
Rabbit
Protein A
RUO
14208, 259229, 5496
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.