missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1J Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35276-100ul
This item is not returnable.
View return policy
Description
PPM1J Polyclonal antibody specifically detects PPM1J in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PPM1J | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| DKFZp434P1514, FLJ35951, MGC19531, MGC90149, PP2CZ, PP2Czeta, PP2C-zeta, PPP2CZEC 3.1.3.16, protein phosphatase 1J, protein phosphatase 1J (PP2C domain containing), protein phosphatase 2a, catalytic subunit, zeta isoform, Protein phosphatase 2C isoform zeta, protein phosphatase 2C zeta, protein phosphatase, Mg2+/Mn2+ dependent, 1J | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-260 of human PPM1J (NP_005158.5).,, Sequence:, VEGRRSVTGVPREPSRGQGLCFYYWGLFDGHAGGGAAEMASRLLHRHIREQLKDLVEILQDPSPPPLCLPTTPGTPDSSDPSHLLGPQSCWSSQKEVSHESLVVGAVENAFQLMDEQMARERRGHQVEGGC | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 333926 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction