missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM1K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31444
This item is not returnable.
View return policy
Description
PPM1K Polyclonal specifically detects PPM1K in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| PPM1K | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| PPM1K | |
| Recombinant protein encompassing a sequence within the center region of human PPM1K. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| DKFZp667B084, DKFZp761G058, mitochondrial, PP2C domain-containing protein phosphatase 1K, PP2Ckappa, PP2C-like mitochondrial protein, PP2Cm, PP2C-type mitochondrial phosphoprotein phosphatase, protein phosphatase 2C kappa, protein phosphatase, Mg2+/Mn2+ dependent, 1K, PTMPEC 3.1.3.16 | |
| Rabbit | |
| 41 kDa | |
| 100 μL | |
| Protein Phosphatase | |
| 152926 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction