Learn More
PPM1N Polyclonal Antibody
GREENER_CHOICE
PPM1N Polyclonal Antibody
Brand:  Invitrogen PA569804
Product Code. 15848213
  • € 530.00 / 100µL
Description
PPM1N Polyclonal Antibody for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PPM1N
Polyclonal
Unconjugated
PPM1N
PPM1N; Probable protein phosphatase 1N; protein phosphatase, Mg2+/Mn2+ dependent 1N (putative); protein phosphatase, Mg2+/Mn2+ dependent, 1N (putative)
Rabbit
Affinity Chromatography
RUO
147699
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q8N819
PPM1N
synthetic peptide directed towards the N-terminal region of human PPM1N.