missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPM2C Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21176-1-AP-20UL
This item is not returnable.
View return policy
Description
PDP1 is a Pyruvate dehydrogenase and helps catalyze phosphorylation of serine residues of E1 to inactivate the E1 component and inhibit the complex. Pyruvate dehydrogenase phosphatases catalyze the dephosphorylation and activation of the E1 component to reverse the effects of pyruvate dehydrogenase kinases. Pyruvate dehydrogenase phosphatase is a heterodimer consisting of catalytic and regulatory subunits.Specifications
| PPM2C | |
| Polyclonal | |
| Unconjugated | |
| PDP1 | |
| PDH, PDP, PDP 1, PDP1, PDPC, PDPC 1, PPM2C, Protein phosphatase 2C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 381511, 54704 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q3UV70, Q9P0J1 | |
| Pdp1 | |
| PPM2C Fusion Protein Ag15453 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction