missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1CB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55136-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to PPP1CB.
SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.Specifications
| PPP1CB | |
| Polyclonal | |
| Unconjugated | |
| PPP1CB | |
| PP1 Beta, PP 1B, PPP1CB, PPP1CD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 19046, 25594, 5500 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.65 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P62140, P62141 | |
| Ppp1cb | |
| Peptide | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction