Learn More
PPP1R12C Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25157-1-AP-20UL
Description
The PPP1R12C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PPP1R12C. This antibody recognizes human, mouse, rat antigen. The PPP1R12C antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PPP1R12C | |
| 280 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| LENG3, MBS85, p84, PPP1R12C | |
| 20 μL | |
| 54776 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |