Learn More
PPP1R12C Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25157-1-AP-150UL
 View more versions of this product
Product Code. 18066685
  • € 410.00 / 150µL
Description
The PPP1R12C antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PPP1R12C. This antibody recognizes human, mouse, rat antigen. The PPP1R12C antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
PPP1R12C
280 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, ELISA
LENG3, MBS85, p84, PPP1R12C
150 μL
54776
Store at -20°C. Stable for one year after shipment.