missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PPP1R14B Polyclonal antibody specifically detects PPP1R14B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | PPP1R14B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | PHI-1, PLCB3Nphospholipase C beta-3 neighboring gene protein, PNGPhospholipase C-beta-3 neighbouring gene protein, protein phosphatase 1 regulatory subunit 14B, protein phosphatase 1, regulatory (inhibitor) subunit 14B, SOM172 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 68-147 of human PPP1R14B (NP_619634.1). RLNLEEWILEQLTRLYDCQEEEIPELEIDVDELLDMESDDARAARVKELLVDCYKPTEAFISGLLDKIRGMQKLSTPQKK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?