missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PPP1R14C Polyclonal Antibody

Product Code. 15653785
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15653785 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15653785 Supplier Invitrogen™ Supplier No. PA549854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PP1R14C protein.

Inhibitor of the PP1 regulatory subunit PPP1CA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPP1R14C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPP1R14C
Gene Accession No. Q8R4R9, Q8R4S0, Q8TAE6
Gene Alias 6330514J04Rik; CPI17-like; Kepi; kinase C-enhanced PP1 inhibitor; kinase-enhanced PP1 inhibitor; NY-BR-81; PKC-potentiated PP1 inhibitory protein; Ppp1r14c; protein phosphatase 1 regulatory inhibitor subunit 14C; protein phosphatase 1 regulatory subunit 14C; protein phosphatase 1, regulatory (inhibitor) subunit 14C; serologically defined breast cancer antigen NY-BR-81
Gene Symbols PPP1R14C
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human PP14C (aa 51-100).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171010, 76142, 81706
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.