Learn More
PPP1R15A Mouse anti-Human, Mouse, Clone: OTI2B11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504309
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The induction of this gene by ionizing radiation occurs in certain cell lines regardless of p53 status, and its protein response is correlated with apoptosis following ionizing radiation.Specifications
| PPP1R15A | |
| Monoclonal | |
| Unconjugated | |
| PPP1R15A | |
| GADD34 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 17872, 23645 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2B11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O75807, P17564 | |
| PPP1R15A | |
| Full length human recombit protein of human PPP1R15A produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.