missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R18 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97370AF488
This item is not returnable.
View return policy
Description
PPP1R18 Polyclonal specifically detects PPP1R18 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PPP1R18 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 170954 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| HKMT1098, KIAA1949, phostensin, PPP1R18, Protein Phosphatase 1 F-Actin Cytoskeleton Targeting Subunit, Protein Phosphatase 1 F-Actin Cytoskeleton-Targeting Subunit, Protein Phosphatase 1 Regulatory Subunit 18, Protein Phosphatase 1, Regulatory Subunit 18 | |
| Produced in rabbits immunized with E. coli-derived Human PPP1R18 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction