Learn More
PPP1R18 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-18432AF647
Product Code. 18546617
  • € 658.00 / 0.10mL
Description
PPP1R18 Polyclonal antibody specifically detects PPP1R18 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PPP1R18
Polyclonal
Western Blot
HKMT1098, KIAA1949, phostensin, PPP1R18, Protein Phosphatase 1 F-Actin Cytoskeleton Targeting Subunit, Protein Phosphatase 1 F-Actin Cytoskeleton-Targeting Subunit, Protein Phosphatase 1 Regulatory Subunit 18, Protein Phosphatase 1, Regulatory Subunit 18
Recognizes region between residues Between Between 325 and 375..
0.1 mL
Primary
Human
Purified
Western Blot
Alexa Fluor 647
50mM Sodium Borate
Rabbit
Affinity purified
RUO
170954
Store at 4°C in the dark.