missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R1A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102875
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PPP1R1A.
Protein phosphatase inhibitor-1 (also designated inhibitor of protein phospha-tase 1, IPP-1 and I-1) plays a role in regulating the phosphorylation of other proteins, and is itself phosphorylated by a cyclic AMP-dependent protein kinase at Threonine 35. In addition, the proline-directed kinases Cdk1, Cdk5, and mitogen-activated protein kinase (MAPK) mediate in vitro phosphorylation of IPP-1 at the phylogenetically conserved position Serine 67. In striatal tissues, glutamate-dependent regulation of N-methyl-d-aspartic acid-type channels influences IPP-1 phosphorylation at Ser 67. The localization and expression of IPP-1 suggests that it may play discrete roles in certain regions and developing stages of the brain, independent of the regulation of protein phosphatase type 1 (PP-1). PP-1 binds to both phosphorylated and dephosphorylated IPP-1. Conversion of PP-1 to a Mn++-dependent state appears to play a role in its regulation by IPP-1. IPP-1 attenuates the activity of glycogen phosphorylase and is thought to be important in the hormonal control of glycogen metabolism.
Specifications
| PPP1R1A | |
| Polyclonal | |
| Unconjugated | |
| Ppp1r1a | |
| 0610038N18Rik; I1; I-1; inhibitor-1; Ipp1; IPP-1; PP1 inhibitor 1; Ppp1r1a; protein phosphatase 1 regulatory inhibitor subunit 1A; protein phosphatase 1 regulatory subunit 1A; protein phosphatase 1, regulatory (inhibitor) subunit 1A; protein phosphatase inhibitor 1; protein phosphatase inhibitor-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5502 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13522 | |
| Ppp1r1a | |
| A synthesized peptide derived from human PPP1R1A(Accession Q13522), corresponding to amino acid residues G128-V171. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction