missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R1B, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00084152-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MDPKDRKKIQFSVPAPPSQLDPRQVEMIRRRRPTPAMLFRLSEHSSPEEEASPHQRASGEGHHLKSKRPNPCAYTPPSLKAVQRIAESHLQSISNLNENQASEEEDELGELRELGYPREEDEEEEEDDEEEEEEEDSQAEVLKVIRQSAGQKTTCGQGLEGPWERPPPLDESERDGGSEDQVEDPALSEPGEEPQRPSPSEPGTSpecifications
| PPP1R1B | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_032192.2 | |
| PPP1R1B | |
| PPP1R1B (NP_115568.2, 1 a.a. ∼ 204 a.a) full-length human protein. | |
| RUO | |
| 84152 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| protein phosphatase 1, regulatory (inhibitor) subunit 1B | |
| DARPP-32/DARPP32/FLJ20940 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction