missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561787
This item is not returnable.
View return policy
Description
Immunogen sequence: YLISLGADRD ATNDDGDLPS DLIDPDYKEL VELFKGTTMD Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 93%.
Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
Specifications
| PPP1R27 | |
| Polyclonal | |
| Unconjugated | |
| PPP1R27 | |
| dysferlin interacting protein 1; Dysferlin-interacting protein 1; dysferlin-interacting protein 1 (toonin); DYSFIP1; PPP1R27; Protein phosphatase 1 regulatory subunit 27; protein phosphatase 1, regulatory subunit 27; toonin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116729 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86WC6 | |
| PPP1R27 | |
| Recombinant protein fragment of Human PPP1R27 (Product #RP-98932) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction