missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118183
This item is not returnable.
View return policy
Description
PPP1R27 Polyclonal Antibody for Western Blot
Inhibits phosphatase activity of protein phosphatase 1 (PP1) complexes.
Specifications
| PPP1R27 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q86WC6 | |
| PPP1R27 | |
| E. coli-derived Human PPP1R27 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PPP1R27 | |
| dysferlin interacting protein 1; Dysferlin-interacting protein 1; dysferlin-interacting protein 1 (toonin); DYSFIP1; PPP1R27; Protein phosphatase 1 regulatory subunit 27; protein phosphatase 1, regulatory subunit 27; toonin | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 116729 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction