missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21334-25ul
This item is not returnable.
View return policy
Description
PPP1R3A Polyclonal antibody specifically detects PPP1R3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PPP1R3A | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| glycogen and sarcoplasmic reticulum binding subunit, skeletal muscle, GM, PP1G, PPP1R3, protein phosphatase 1 glycogen- associated regulatory subunit, Protein phosphatase 1 glycogen-associated regulatory subunit, protein phosphatase 1 regulatory subunit 3A, protein phosphatase 1, regulatory (inhibitor) subunit 3A, protein phosphatase 1, regulatory (inhibitor) subunit 3A (glycogen andsarcoplasmic reticulum binding subunit, skeletal muscle), Protein phosphatase type-1 glycogen targeting subunit, RG1, serine/threonine specific protein phosphatase, type-1 protein phosphatase skeletal muscle glycogen targeting subunit | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: CNSTREIQGIEKHPYPESKPEEVSRSSGIVTSGSRKERCIGQIFQTEEYSVEKSLGPMILINKPLENMEEARHENEGLVSSGQSLYTSGEKESDSSASTSLPVEESQAQGNESLFS | |
| 25 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5506 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction