missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32351
This item is not returnable.
View return policy
Description
PPP1R3E Polyclonal specifically detects PPP1R3E in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PPP1R3E | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Protein Phosphatase 1 Regulatory Subunit 3E, Protein Phosphatase 1, Regulatory (Inhibitor) Subunit 3E, Protein Phosphatase 1, Regulatory Subunit 3E | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 90673 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| PPP1R3E | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MSRERPPGTDIPRNLSFIAALTERAYYRSQRPSLEEEPEEEP | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction