missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R3F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12626-50ul
This item is not returnable.
View return policy
Description
PPP1R3F Polyclonal antibody specifically detects PPP1R3F in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| PPP1R3F | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| HB2E, protein phosphatase 1 regulatory subunit 3F, protein phosphatase 1, regulatory subunit 3F, regulatory (inhibitor) subunit 3F | |
| Produced in rabbits immunized with E. coli-derived Human PPP1R3F fragment | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 89801 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction