missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R3F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118184
This item is not returnable.
View return policy
Description
PPP1R3F Polyclonal Antibody for Western Blot, IP
Glycogen-targeting subunit for protein phosphatase 1 (PP1).
Specifications
| PPP1R3F | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q6ZSY5 | |
| PPP1R3F | |
| E. coli-derived Human PPP1R3F fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PPP1R3F | |
| DXImx48e; HB2E; LL0XNC01-7P3.1; Ppp1r3f; Protein phosphatase 1 regulatory subunit 3F; protein phosphatase 1, regulatory (inhibitor) subunit 3F; protein phosphatase 1, regulatory subunit 3F; R3F; RF3; Sfc15 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 89801 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction