missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R7 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97835FR
This item is not returnable.
View return policy
Description
PPP1R7 Polyclonal antibody specifically detects PPP1R7 in Human samples. It is validated for Western Blot
Specifications
| PPP1R7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| Protein phosphatase 1 regulatory subunit 22, protein phosphatase 1 regulatory subunit 7, protein phosphatase 1, regulatory (inhibitor) subunit 7, protein phosphatase-1 regulatory subunit 7 alpha2, protein phosphatase-1 regulatory subunit 7 beta1, protein phosphatase-1 regulatory subunit 7 beta2, regulatory subunit 7, sds22 | |
| Produced in rabbits immunized with E. coli-derived Human PPP1R7 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 5510 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction