missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R8 Monoclonal Antibody (OTI1E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804570
This item is not returnable.
View return policy
Description
This gene, through alternative splicing, encodes three different isoforms. Two of the protein isoforms encoded by this gene are specific inhibitors of type 1 serine/threonine protein phosphatases and can bind but not cleave RNA. The third protein isoform lacks the phosphatase inhibitory function but is a single-strand endoribonuclease comparable to RNase E of E. coli. This isoform requires magnesium for its function and cleaves specific sites in A+U-rich regions of RNA.Specifications
| PPP1R8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12972 | |
| PPP1R8 | |
| Human recombit protein fragment corresponding to amino acids 1-225 of human PPP1R8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1E2 | |
| Unconjugated | |
| PPP1R8 | |
| ARD-1, ARD1, NIPP-1, NIPP1, PRO2047 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5511 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction