missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPP1R8 Monoclonal Antibody (OTI1E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804570S
This item is not returnable.
View return policy
Description
PPP1R8 Monoclonal antibody specifically detects PPP1R8 in Human samples. It is validated for Western BlotSpecifications
| PPP1R8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17931 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3958 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1E2 | |
| Unconjugated | |
| LGALS3 | |
| 6330548N22Rik; activator of RNA decay; ARD1; ARD-1; AU044684; LOC100722083; NIPP1; NIPP-1; NIPP-1, nuclear inhibitor of protein phosphatase-1; Nuclear inhibitor of protein phosphatase 1; nuclear inhibitor of protein phosphatase-1 alpha; nuclear inhibitor of protein phosphatase-1 beta; nuclear subunit of PP-1; Ppp1r8; PRO2047; protein phosphatase 1 regulatory inhibitor subunit 8; protein phosphatase 1 regulatory subunit 8; protein phosphatase 1, regulatory (inhibitor) subunit 8; protein phosphatase 1, regulatory inhibitor subunit 8; protein phosphatase 1, regulatory subunit 8; RNase E | |
| Full length human recombinant protein of human LGALS3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction