Learn More
PPP1R8 Mouse anti-Human, Clone: OTI5H9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804813
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene, through alternative splicing, encodes three different isoforms. Two of the protein isoforms encoded by this gene are specific inhibitors of type 1 serine/threonine protein phosphatases and can bind but not cleave RNA. The third protein isoform lacks the phosphatase inhibitory function but is a single-strand endoribonuclease comparable to RNase E of E. coli. This isoform requires magnesium for its function and cleaves specific sites in A+U-rich regions of RNA.Specifications
| PPP1R8 | |
| Monoclonal | |
| Unconjugated | |
| PPP1R8 | |
| ARD-1, ARD1, NIPP-1, NIPP1, PRO2047 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5511 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| OTI5H9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q12972 | |
| PPP1R8 | |
| Human recombit protein fragment corresponding to amino acids 1-225 of human PPP1R8 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.