missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PPP1R9A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110944
This item is not returnable.
View return policy
Description
Immunogen sequence: EEPCAESKAM PKSEIPSPQS QLLEDAEANL VGREAAKQQR KELAGGDFTS PDASASSCGK EVPEDSNNFD GSHVYMHSDY NVYRVRSRYN SDWGETGTEQ DEEEDSDENS YYQP Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 77%.
Neurabin I is a brain-specific protein which contains an F-actin binding domain, a PDZ domain, a transmembrane-protein-interacting-domain, and a coiled-coil region. As its structure suggests, neurabin I is a promiscuous protein binding to F-actin, protein phosphatase 1, TGN38, and p70 S6 kinase. Found exclusively in brain, this protein is highly concentrated in the synapse and has also been shown to be enriched in the lamellipodia of the growth cone during neuronal development. Neurabin I appears to be a bridging protein by targeting other proteins to the synapse or by linking membrane proteins to the actin cytoskeleton.
Specifications
| PPP1R9A | |
| Polyclonal | |
| Unconjugated | |
| PPP1R9A | |
| 2810430P21Rik; 4930518N04Rik; 5330407E15; A230094E16Rik; actin-binding protein neurabin; BB181831; KIAA1222; mKIAA1222; Neb1; neurabin 1; Neurabin I; neurabin-1; Neurabin-I; neural tissue-specific F-actin binding protein; neural tissue-specific F-actin-binding protein I; NRB; NRB1; NRBI; p180; PP1bp175; Ppp1r9a; Protein phosphatase 1 regulatory subunit 9A; protein phosphatase 1, regulatory (inhibitor) subunit 9A; protein phosphatase 1, regulatory subunit 9A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55607 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULJ8 | |
| PPP1R9A | |
| Recombinant Protein corresonding to Human PPP1R9A. Recombinant protein control fragment (Product #RP-92313). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction