missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Antibody (SOX2/3169R), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08508PE
This item is not returnable.
View return policy
Description
SOX2 Monoclonal antibody specifically detects SOX2 in Human, Mouse, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifications
| SOX2 | |
| Monoclonal | |
| PE | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| Recombinant fragment (within aa176-305) of human SOX2 protein (exact sequence is proprietary) (Uniprot: P48431) | |
| 0.1 mL | |
| Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 6657 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| SOX2/3169R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction